Learn More
Gilson™ ASPEC™ WCX (CARB AC) SPE 96-well Plate
Silica based WCX cartridges, contain a high-quality, weak cation exchange sorbent (carboxylic acid), for extracting strong basic compounds (pKa > 9) from aqueous and organic solutions
sur devis
Spécification
Volume (métrique) 2 ml
Consulter d'autres spécifications
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
2 ml