Learn More
Gilson™ ASPEC™ WCX SPE Cartridge, Bed Wt. 200 mg
Silica based WCX cartridges, contain a high-quality, weak cation exchange sorbent (carboxylic acid), for extracting strong basic compounds (pKa > 9) from aqueous and organic solutions
Marque:  Gilson™ 54350557
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646285
  • 193.00€ / 50 pièces
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
WCX
Automated extractions
200 mg
WCX
ASPEC™
3 mL