Learn More
Gilson™ ASPEC™ X-SAX SPE Cartridge, Bed Wt. 500 mg
Polymer based X-SAX cartridges contain a high-quality, polystyrene-divinylbenzene copolymer functionalized by a strong anion exchanger, for extracting acidic drugs and metabolites (pKa 2 to 8) from biological fluids and tissues
Marque:  Gilson™ 54350865
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16626635
  • 400.00€ / 30 pièces
Date d’expédition estimée: 19-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
X-SAX
Automated extractions
500 mg
X-SAX
ASPEC™
6 mL