Learn More
Gilson™ ASPEC™ X-SCX SPE Cartridge, Bed Wt. 500 mg
Polymer based X-SCX cartridges contain a high-quality, polystyrene-divinylbenzene copolymer functionalized by a strong cation exchanger, for extracting basic drugs and metabolites (pKa 7 to 9) from biological fluids and tissues
Marque:  Gilson™ 54350859NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16616625
  • 361.00€ / 30 pièces
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
X-SCX
Manual extractions
500 mg
X-SCX
ASPEC™
6 mL