Learn More
Gilson™ ASPEC™ X-WAX SPE Cartridge, Bed Wt. 100 mg
Polymer based X-WAX cartridges contain a high-quality, polystyrene-divinylbenzene copolymer functionalized by a weak anion exchanger (amine), for catching and releasing strong acidic compounds (pKa < 3) when extracting strong acidic drugs and metabolites from biological fluids
Marque:  Gilson™ 54350875
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16626655
  • 224.00€ / 30 pièces
Date d’expédition estimée: 24-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
Yes
SPE
X-WAX
Automated extractions
100 mg
X-WAX
ASPEC™
6 mL