Learn More
Gilson™ ASPEC™ X-WAX SPE Cartridge, Bed Wt. 60 mg
Polymer based X-WAX cartridges contain a high-quality, polystyrene-divinylbenzene copolymer functionalized by a weak anion exchanger (amine), for catching and releasing strong acidic compounds (pKa < 3) when extracting strong acidic drugs and metabolites from biological fluids
Marque:  Gilson™ 54350874NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16616655
  • 212.00€ / 50 pièces
Date d’expédition estimée: 24-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
X-WAX
Manual extractions
60 mg
X-WAX
ASPEC™
3 mL