Learn More
Gilson™ ASPEC™ X-WCX SPE 96-well Plate
Polymer based X-WCX cartridges contain a high-quality, polystyrene-divinylbenzene copolymer functionalized by a weak cation exchanger, for catching and releasing strong basic compounds (pKa > 10) during extraction from biological fluids
Marque:  Gilson™ 54350935NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16646715
  • 346.00€ / 1 pièce
Date d’expédition estimée: 23-11-2021
Ajouter au panier
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
SPE 96-Well Plate
No
SPE
X-WCX
10 mg
X-WCX
ASPEC™
2 mL