Learn More
Gilson™ ASPEC™ X-WCX SPE 96-well Plate
Polymer based X-WCX cartridges contain a high-quality, polystyrene-divinylbenzene copolymer functionalized by a weak cation exchanger, for catching and releasing strong basic compounds (pKa > 10) during extraction from biological fluids
sur devis
Spécification
Volume (métrique) 2 ml
Consulter d'autres spécifications
Description
Description
  • ASPEC™ pre-capped cartridges are ASPEC™ instrument-ready, saving your time and money by eliminating manual capping.
  • Provide high loading capacity, accurate elution properties and reproducible separation with low solvent usage
  • Available uncapped for manual extractions
Spécification
Spécification
2 ml