Learn More
Gilson™ ASPEC™ X-WCX SPE Cartridge, Bed Wt. 100 mg
Polymer based X-WCX cartridges contain a high-quality, polystyrene-divinylbenzene copolymer functionalized by a weak cation exchanger, for catching and releasing strong basic compounds (pKa > 10) during extraction from biological fluids
Marque:  Gilson™ 54350869NC
Informations supplémentaires : Poids : 0.01100kg
 Afficher plus de versions de ce produit
Code produit. 16616645
  • 200.00€ / 30 pièces
Date d’expédition estimée: 22-11-2021
Ajouter au panier
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Spécification
Spécification
SPE cartridge
No
SPE
X-WCX
Manual extractions
100 mg
X-WCX
ASPEC™
6 mL