missing translation for 'onlineSavingsMsg'
Learn More

ATP6V0A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 13828-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16802224

  • 399.00€ / 150µL
Expédition estimée: 12-04-2024
pour voir le stock disponible 

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

ATP6V0A1 encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c′, c′, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This gene encodes one of three A subunit proteins and the encoded protein is associated with clathrin-coated vesicles.
Spécification

Spécification

ATP6V0A1
Polyclonal
Unconjugated
ATP6V0A1
a1, ATP6N1, ATP6N1A, ATP6V0A1, Stv1, V ATPase 116 kDa isoform a1, Vacuolar proton pump subunit 1, Vph1, VPP1
Rabbit
Antigen Affinity Chromatography
RUO
11975, 29757, 535
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P25286, Q93050, Q9Z1G4
ATP6V0A1
ATP6V0A1 Fusion Protein Ag4191
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Suggestions de produits

Suggestions de produits

Vidéos
FDS
Documents

Documents

Certificats
Promotions

Promotions