missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ATP6V1B1 (Human) Recombinant Protein

Human ATP6V1B1 full-length ORF ( AAH63411, 1 a.a. - 513 a.a.) recombinant protein with GST-tag at N-terminal.

Marque:  Abnova™ H00000525-P01.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16122071

  • 310.00€ / 10µg

Expédition estimée
21-12-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

  • Sequence: MAMEIDSRPGGLPGSSCNLGAAREHMQAVTRNYITHPRVTYRTVCSVNGPLVVLDRVKFAQYAEIVHFTLPDGTQRSGQVLEVAGTKAIVQVFEGTSGIDARKTTCEFTGDILRTPVSEDMLGRVFNGSGKPIDKGPVVMAEDFLDINGQPINPHSRIYPEEMIQTGISPIDVMNSIARGQKIPIFSAAGLPHNEIAAQICRQAGLVKKSKAVLDYHDDNFAIVFAAMGVNMETARFFKSDFEQNGTMGNVCLFLNLANDPTIERIITPRLALTTAEFLAYQCEKHVLVILTDMSSYAEALREVSAAREEVPGRRGFPGYMYTDLATIYERAGRVEGRGGSITQIPILTMPNDDITHPIPDLTGFITEGQIYVDRQLHNRQIYPPINVLPSLSRLMKSAIGEGMTRKDHGDVSNQLYACYAIGKDVQAMKAVVGEEALTSEDLLYLEFLQKFEKNFINQGPYENRSVFESLDLGWKLLRIFPKEMLKRIPQAVIDEFYSREGALQDLAPDTAL
Spécification

Spécification

AAH63411
ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATP6V1B1
GST
525
Wheat germ expression system
10 μg
ATP6B1, MGC32642, RTA1B, VATB, VMA2, VPP3
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Vidéos
FDS
Documents