missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATXN2L Rabbit anti-Human, Mouse, Proteintech
Rabbit PolyClonal Antibody
Marque: Proteintech 24822-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes an ataxin type 2 related protein of unknown function. This protein is a member of the spinocerebellar ataxia (SCAs) family, which is associated with a complex group of neurodegenerative disorders. Several alternatively spliced transcripts encoding different isoforms have been found for this gene.Spécification
ATXN2L | |
Polyclonal | |
Unconjugated | |
ATXN2L | |
A2D, A2LG, A2LP, A2RP, Ataxin 2 domain protein, ataxin 2 like, Ataxin 2 like protein, Ataxin 2 related protein, ATXN2L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11273, 233871 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7TQH0, Q8WWM7 | |
Atxn2l | |
ATXN2L Fusion Protein Ag20581 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |