missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AVPR1B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | AVPR1B |
---|---|
Concentration | 0.2 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16816615
|
Proteintech
55142-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 03-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16806615
|
Proteintech
55142-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 03-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Vasopressin V1B receptor expression has been documented in the pituitary, pituitary tumors, pheochromocytoma, breast cancer, and small-cell lung carcinomas. G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more than a thousand members currently predicted to exist. All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates. Two blockbuster drugs which have been developed as GPCR-targeted pharmaceuticals are Zyprexa (Eli Lilly) and Claritin (Schering-Plough) which have multi-billion dollar shares of the mental health and allergy markets, respectively.Spécification
AVPR1B | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
P47901 | |
553 | |
Peptide | |
Primary | |
-20°C | |
AVPR1B |
0.2 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
AVPR V1b, AVPR V3, AVPR1B, AVPR3, V1bR, Vasopressin V1b receptor, Vasopressin V3 receptor, VPR3 | |
AVPR1B | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |