missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BACE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | BACE2 |
---|---|
Concentration | 0.13 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16845914
|
Proteintech
16321-1-AP-20UL |
20 μL |
140.00€
20µL |
Expédition estimée: 13-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
16835914
|
Proteintech
16321-1-AP-150UL |
150 μL |
399.00€
150µL |
Expédition estimée: 13-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Cerebral deposition of amyloid beta peptide is an early and critical feature of Alzheimer′s disease and a frequent complication of Down syndrome. Amyloid beta peptide is generated by proteolytic cleavage of amyloid precursor protein by 2 proteases, one of which is the protein encoded by this gene. This gene localizes to the ′Down critical region′ of chromosome 21. The encoded protein, a member of the peptidase A1 protein family, is a type I integral membrane glycoprotein and aspartic protease. Three transcript variants encoding different isoforms have been described for this gene.Spécification
BACE2 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q6IE75, Q9JL18, Q9Y5Z0 | |
25825, 288227, 56175 | |
BACE2 Fusion Protein Ag9404 | |
Primary | |
-20°C | |
BACE2 |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AEPLC, ALP56, Asp 1, ASP1, ASP21, Aspartic like protease 56 kDa, Aspartyl protease 1, BACE2, BAE2, Beta secretase 2, CDA13, CEAP1, Down region aspartic protease, DRAP, Memapsin 1, Theta secretase, UNQ418/PRO852 | |
Bace2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |