missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BAG6 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26417-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a protein with two leucine zipper domains, a ribosomal protein S14 signature domain and a Sec39 like domain. The protein is thought to be involved in Golgi-to-ER transport. Mutations in this gene are associated with short stature, optic nerve atrophy, and Pelger-Huet anomaly. Involved in Golgi-to-endoplasmic reticulum (ER) retrograde tranport; the function is proposed to depend on its association in the NRZ complex which is believed to play a role in SNARE assembly at the ER.Spécification
BAG6 | |
Polyclonal | |
Unconjugated | |
BAG6 | |
BAG 6, BAG6, BAT3, D6S52E, G3, HLA B associated transcript 3, Protein G3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
7917 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P46379 | |
BAG6 | |
BAG6 Fusion Protein Ag24525 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |