Learn More
Fisherbrand™ BagPage™ Full Page Blender Bags
A blender bag with full-page micro-perforated filter, available in two sizes.
196.00€
Spécification
Type de capuchon Heat seal or clip
À utiliser avec (application) Ideal for homogenizing pasty samples
Type de produit Full-page Bag
Stérilité Sterile
Couleur Clear
Consulter d'autres spécifications
Produits 1
Code produit Marque Capacité (métrique) Prix Quantité et disponibilité  
Code produit Marque Capacité (métrique) Prix Quantité et disponibilité  
10010051 Afficher les documents Fisherbrand™
100mL 196.00€
500 pièces View alternative products
missing translation for 'toSaveUpto' 
»
Expédition estimée
04-05-2020
Add to basket
Description
Description
  • Ideal for homogenizing pasty samples
  • Multilayer, reinforced multicoated complex
  • Full-surface micro-perforated filter with porosity of 280 microns
  • Rigid and transparent construction
  • Filtration during homogenization
  • No need to wait for debris sedimentation
  • Particle-free solution, precise results, and excellent reproducibility
  • Gamma ray treated
  • Same bag is used to homogenize, filter and pipet
  • No contact between the sample and the blender
  • Can be closed with a clip or heat-sealed
  • Compatible with any lab blender
  • Approved for food contact
Specifications
Specifications
Heat seal or clip
Full-page Bag
Clear
Ideal for homogenizing pasty samples
Sterile