missing translation for 'onlineSavingsMsg'
Learn More

BD Recombinant Human IFN-γ

Pure by SDS-PAGE, and an absorbance assay based on the Beers-Lambert law

Marque:  BD 554616

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15867808

Quantité Prix
1 576.30€ / 1 pièce
6 537.16€ / 1 pièce
Date d’expédition estimée: 23-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!



Description

Description

Frozen liquid comprised of 0.22μm sterile-filtered aqueous buffered solution and containing 2.5mg/ml biotechnology grade, low endotoxin bovine serum albumin, with no azide

  • Reactivity: Human
  • Storage Buffer: Frozen aqueous buffered solution containing BSA
  • Biological Activity: Activity range is measured in an anti-viral bioassay using A549 indicator cells
  • Specific Activity: 0.1 - 2.0 x 108 Units/mg
  • ED50: 0.05 to 1.0 ng/ml
  • Observed dose response relationship: 16X

Bioassay, enzyme-linked immunosorbent assay standard

Spécification

Spécification

250μg/mL
Solution
14 to 18
SDS-PAGE
-80°C
Yes
Bioassay, ELISA Standard
Frozen aqueous Buffered solution containing BSA
Human IFN-gamma Recombinant Protein
25μg
Human
Vidéos
FDS
Documents
Certificats
Promotions

Promotions

Research Use Only