missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BET1L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14163-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Vesicle SNARE required for targeting and fusion of retrograde transport vesicles with the Golgi complex. Required for the integrity of the Golgi complex.Spécification
BET1L | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35152, O35153, Q9NYM9 | |
BET1 like protein, BET1L, BET1L1, GOLIM3, GOS 15, GS15, HSPC197 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
51272, 54399, 54400 |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
BET1L | |
Liquid | |
BET1L | |
BET1L Fusion Protein Ag5351 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |