missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BID (Human) Recombinant Protein
Human BID full-length ORF ( AAH09197, 1 a.a. - 195 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00000637-P01.10ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Sequence: MDCEVNNGSSLRDECITNLLVFGFLQSCSDNSFRRELDALGHELPVLAPQWEGYDELQTDGNRSSHSRLGRIEADSESQEDIIRNIARHLAQVGDSMDRSIPPGLVNGLALQLRNTSRSEEDRNRDLATALEQLLQAYPRDMEKEKTMLVLALLLAKKVASHTPSLLRDVFHTTVNFINQNLRTYVRSLARNGMD
Spécification
AAH09197 | |
BH3 interacting domain death agonist | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BID | |
GST |
637 | |
Wheat germ expression system | |
10 μg | |
FP497, MGC15319, MGC42355 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |