Learn More
ACROBiosystems Biotinylated Cynomolgus / Rhesus macaque FcRn Heterodimer Protein, His, Avitag™&Strep II Tag (BLI verified)
Biotinylated Protein
423.38€ - 1862.87€
Spécification
Numéro d’adhésion | NP_001244449.1 (FCGRT) and Q8SPW0.1 (B2M) |
---|---|
Poids moléculaire | 34.1 kDa (FCGRT) and 13.0 kDa (B2M) |
Source | MABSol™ Biotinylated Cynomolgus / Rhesus macaque FcRn Heterodimer Protein, His,AviTag&Strep; II Tag (BLI verified) (FCM-C82W5) is expressed from human HEK293 cells. It contains AA Ala 24 - Ser 297 (FCGRT) and Ile 21 - Met 119 (B2M) (Accession # Q8SPV9-1 (FCGRT) and Q8SPW0-1 (B2M)). In the region Ala 24 - Ser 297 (FCGRT) and Ile 21 - Met 119 (B2M), the AA sequence of Cynomolgus and Rhesus macaque FcRn (FCGRT and B2M) are homologus. |
Immunogène | Ala 24 - Ser 297 (FCGRT) and Ile 21 - Met 119 (B2M) |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876727
|
ACROBiosystems
FCM-C82W5-25UG |
25 μg |
423.38€
25µg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15866727
|
ACROBiosystems
FCM-C82W5-200UG |
200 μg |
1862.87€
200µg |
En stock Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
FCGRT & B2M heterodimer protein (FcRn complex) consist of two subunits: p51 (equivalent to FCGRT), and p14 (equivalent to beta-2-microglobulin), and forms an MHC class I-like heterodimer. Fc fragment of IgG, receptor, transporter, alpha (FCGRT) binds to the Fc region of monomeric immunoglobulins gamma and mediates the uptake of IgG from milk. FCGRT possible role in transfer of immunoglobulin G from mother to fetus. Beta-2-microglobulin (B2M) is a component of the class I major histocompatibility complex (MHC) and involved in the presentation of peptide antigens to the immune system.
Biotinylated Cynomolgus / Rhesus macaque FcRn Heterodimer Protein, His,Avitag&Strep II Tag (BLI verified), produced by co-expression of FCGRT and B2M, has a calculated MW of 34.1 kDa (FCGRT) and 13.0 kDa (B2M). Subunit FCGRT is fused with an Avi tag (AvitSpecifications
NP_001244449.1 (FCGRT) and Q8SPW0.1 (B2M) | |
MABSol™ Biotinylated Cynomolgus / Rhesus macaque FcRn Heterodimer Protein, His,AviTag&Strep; II Tag (BLI verified) (FCM-C82W5) is expressed from human HEK293 cells. It contains AA Ala 24 - Ser 297 (FCGRT) and Ile 21 - Met 119 (B2M) (Accession # Q8SPV9-1 (FCGRT) and Q8SPW0-1 (B2M)). In the region Ala 24 - Ser 297 (FCGRT) and Ile 21 - Met 119 (B2M), the AA sequence of Cynomolgus and Rhesus macaque FcRn (FCGRT and B2M) are homologus. | |
-20°C | |
FcRn (FCGRT and B2M) | |
Cynomolgus Monkey, Rhesus Macaque | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
34.1 kDa (FCGRT) and 13.0 kDa (B2M) | |
Ala 24 - Ser 297 (FCGRT) and Ile 21 - Met 119 (B2M) | |
FcRn,FCGRT and B2M | |
Precision Avi | |
His Tag and Avi Tag and Strep Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE |