missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human B7-1 / CD80 Protein, Fc, Avitag™

Biotinylated Protein

Marque:  ACROBiosystems B71-H82F2-25UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15875947

  • 353.08€ / 25µg
Date d’expédition estimée: 07-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

B7-1 and B7-2, together with their receptors CD28 and CTLA­4, constitute one of the dominant co-stimulatory pathways that regulate T­ and B­cell responses. Although both CTLA­4 and CD28 can bind to the same ligands, CTLA­4 binds to B7­1 and B7­2 with a 20 ­ 100 fold higher affinity than CD28 and is involved in the down­regulation of the immune response. B-lymphocyte activation antigen B7-1 (referred to as B7) also known as cluster of Differentiation 80 (CD80), is a member of cell surface immunoglobulin superfamily and is expressed on activated B cells, activated T cells, macrophages and dendritic cells. It is the ligand for two different proteins on the T cell surface: CD28 (for autoregulation and intercellular association) and CTLA-4 (for attenuation of regulation and cellular disassociation). CD80 works in tandem with CD86 to prime T cells. CD80 plays a role in induction of innate immune responses by activating NF-κB-signaling pathway in macrophages. CD80 is thus regarded as promising therapeutic targets for autoimmune diseases and various carcinomas.

This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 52.3 kDa. As a result of glycosylation, the protein migrates as 65-95 kDa under reducing (R) condition, and 130-160 kDa under n
Spécification

Spécification

NP_005182.1
HEK293
Human
52.3 kDa
25 µg
Val 35 - Asn 242
CD80,B7,B7-1,B7.1,BB1,CD28LG,CD28LG1,LAB7
Precision Avi
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>95% as determined by SDS-PAGE
Biotinylated Human B7-1, Fc,Avitag (B71-H82F2) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # NP_005182.1).
-20°C
B7-1
Fc Tag and Avi Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions