Learn More
ACROBiosystems Biotinylated Human B7-1 / CD80 Protein, Fc, Avitag™
Biotinylated Protein
447.57€ - 1947.54€
Spécification
Numéro d’adhésion | NP_005182.1 |
---|---|
Poids moléculaire | 52.3 kDa |
Source | Biotinylated Human B7-1, Fc,Avitag (B71-H82F2) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # NP_005182.1). |
Immunogène | Val 35 - Asn 242 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15875947
|
ACROBiosystems
B71-H82F2-25UG |
25 μg |
447.57€
25µg |
Expédition estimée: 29-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15865947
|
ACROBiosystems
B71-H82F2-200UG |
200 μg |
1947.54€
200µg |
Expédition estimée: 29-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
B7-1 and B7-2, together with their receptors CD28 and CTLA-4, constitute one of the dominant co-stimulatory pathways that regulate T- and B-cell responses. Although both CTLA-4 and CD28 can bind to the same ligands, CTLA-4 binds to B7-1 and B7-2 with a 20 - 100 fold higher affinity than CD28 and is involved in the down regulation of the immune response.B-lymphocyte activation antigen B7-1 (referred to as B7) also known as cluster of Differentiation 80 (CD80), is a member of cell surface immunoglobulin superfamily and is expressed on activated B cells, activated T cells, macrophages and dendritic cells. It is the ligand for two different proteins on the T cell surface: CD28 (for autoregulation and intercellular association) and CTLA-4 (for attenuation of regulation and cellular disassociation). CD80 works in tandem with CD86 to prime T cells. CD80 plays a role in induction of innate immune responses by activating NF-κB-signaling pathway in macrophages. CD80 is thus regarded as promising therapeutic targets for autoimmune diseases and various carcinomas.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 52.3 kDa. As a result of glycosylation, the protein migrates as 65-95 kDa under reducing (R) condition, and 130-160 kDa under nSpécification
NP_005182.1 | |
Biotinylated Human B7-1, Fc,Avitag (B71-H82F2) is expressed from human 293 cells (HEK293). It contains AA Val 35 - Asn 242 (Accession # NP_005182.1). | |
-20°C | |
B7-1 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
52.3 kDa | |
Val 35 - Asn 242 | |
CD80,B7,B7-1,B7.1,BB1,CD28LG,CD28LG1,LAB7 | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |