missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Biotinylated Human CD52 Protein, Fc, Avitag™
Biotinylated Protein
423.38€ - 1862.87€
Spécification
Poids moléculaire | 29.7 kDa |
---|---|
Source | Biotinylated Human CD52, Fc,Avitag (CD2-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accession # P31358-1). |
Immunogène | Gly 25 - Ser 36 |
Conditions de stockage | -20°C |
Alias de gène | CDW52,HE5,CD52,CAMPATH-1 antigen |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876157
|
ACROBiosystems
CD2-H82F3-200UG |
200 μg |
1862.87€
200µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15886157
|
ACROBiosystems
CD2-H82F3-25UG |
25 μg |
423.38€
25µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
CAMPATH-1 antigen, also known as cluster of differentiation 52 (CD52), is a glycoprotein that in humans is encoded by the CD52 gene.It is widely expressed on the cell surface of immune cells, such as mature lymphocytes, natural killer cells (NK), eosinophils, neutrophils, monocytes/macrophages, and dendritic cells (DCs).ligation of cell surface CD52 molecules may offer costimulatory signals for T-cell activation and proliferation.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 29.7 kDa. As a result of glycosylation, the protein migrates as 38-45 kDa under reducing (R) condition, and 66-100 kDa under noSpécification
29.7 kDa | |
Gly 25 - Ser 36 | |
CDW52,HE5,CD52,CAMPATH-1 antigen | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
Biotinylated Human CD52, Fc,Avitag (CD2-H82F3) is expressed from human 293 cells (HEK293). It contains AA Gly 25 - Ser 36 (Accession # P31358-1). | |
-20°C | |
CD52 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |