missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Biotinylated Human Glypican 3 / GPC3 Protein, His, Avitag™
Biotinylated Protein
302.42€ - 1318.52€
Spécification
Numéro d’adhésion | NP_004475.1 |
---|---|
Poids moléculaire | 38.1 kDa and 26.4 kDa |
Source | Biotinylated Human Glypican 3, His,Avitag (GP3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1). |
Immunogène | Gln 25 - His 559 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876877
|
ACROBiosystems
GP3-H82E5-200UG |
200 μg |
1318.52€
200µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15886877
|
ACROBiosystems
GP3-H82E5-25UG |
25 μg |
302.42€
25µg |
Expédition estimée: 02-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Glypican-3 (GPC3) is also known as Intestinal protein OCI-5, GTR2-2, MXR7, which belongs to the glypican family. Glypican 3 / GPC-3 is highly expressed in lung, liver and kidney. Glypican-3 inhibits the dipeptidyl peptidase activity of DPP4. Glypican-3 may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and also may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be partially processed into N and C-terminal fragment with calculated MW of 38.1 kDa aSpécification
NP_004475.1 | |
Biotinylated Human Glypican 3, His,Avitag (GP3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1). | |
-20°C | |
Glypican 3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
38.1 kDa and 26.4 kDa | |
Gln 25 - His 559 | |
GPC3,OCI5,Glypican-3,GTR2-2,MXR7,DGSX,SDYS ,SGB,SGBS,SGBS1 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>85% as determined by SDS-PAGE |