Learn More
ACROBiosystems Biotinylated Human Glypican 3 / GPC3 Protein, His, Avitag™
Biotinylated Protein
Marque:  ACROBiosystems GP3-H82E5-200UG
Informations supplémentaires : Poids : 0.00001kg
 Afficher plus de versions de ce produit
Code produit. 15876877
  • 1307.63€ / 200µg
Expédition estimée: 10-04-2024
pour voir le stock disponible 
Description
Description
Glypican-3 (GPC3) is also known as Intestinal protein OCI-5, GTR2-2, MXR7, which belongs to the glypican family. Glypican 3 / GPC-3 is highly expressed in lung, liver and kidney. Glypican-3 inhibits the dipeptidyl peptidase activity of DPP4. Glypican-3 may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and also may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. This protein contains a furin-like convertase cleavage site, 355-RQYR-358, and will be partially processed into N and C-terminal fragment with calculated MW of 38.1 kDa a
Spécification
Spécification
NP_004475.1
200 μg
Gln 25 - His 559
GPC3,OCI5,Glypican-3,GTR2-2,MXR7,DGSX,SDYS ,SGB,SGBS,SGBS1
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>85% as determined by SDS-PAGE
38.1 kDa and 26.4 kDa
Biotinylated Human Glypican 3, His,Avitag (GP3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1).
-20°C
Glypican 3
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins