missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human LILRA1 / CD85i / LIR-6 Protein, His, Avitag™

Biotinylated Protein

Marque:  ACROBiosystems LI1-H82E8-200UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877247

  • 1392.13€ / 200µg
Date d’expédition estimée: 03-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Leukocyte immunoglobulin-like receptors (LILRs) are inhibitory, stimulatory or soluble receptors encoded within the leukocyte receptor complex. Some LILRs are extensively polymorphic, and exhibit evidence for balancing selection and association with disease susceptibility. LILRA1 (LIR-6) can recognize MHC (major histocompatibility complex) class I or class I-like molecules, and high levels of sequence similarity among LILRA1, LILRA2 (ILT1), LILRA3 (ILT6) and LILRB1/B2.

This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 52.2 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

NP 006854.1
HEK293
Human
52.2 kDa
200 µg
Pro 17 - Asn 461
LILRA1,LIR6,CD85i,LIR-6
Precision Avi
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>90% as determined by SDS-PAGE
Biotinylated Human LILRA1, His,Avitag (LI1-H82E8) is expressed from human 293 cells (HEK293). It contains AA Pro 17 - Asn 461 (Accession # O75019-1).
-20°C
LILRA1
His Tag and Avi Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions