missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human LILRB1 / CD85j / ILT2 Protein, Fc, Avitag™

Biotinylated Protein

Marque:  ACROBiosystems CDJ-H82F7-200UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15876407

  • 1392.13€ / 200µg
Date d’expédition estimée: 03-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

CD85 antigen-like family member J (CD85J) is also known as Leukocyte immunoglobulin-like receptor subfamily B member 1 (LILRB1 or LIR-1), Immunoglobulin-like transcript 2 (ILT-2), Monocyte/macrophage immunoglobulin-like receptor 7 (MIR7), which belongs to leukocyte immunoglobulin-like receptor (LIR) family. CD85J / LILRB1 Contains 4 Ig-like C2-type (immunoglobulin-like) domains. CD85J / LIR-1 is expressed predominantly on B-cells and monocytes. CD85J is receptor for class I MHC antigens and recognizes a broad spectrum of HLA-A, HLA-B, HLA-C and HLA-G alleles and is also receptor for H301/UL18, a human cytomegalovirus class I MHC homolog. Ligand binding results in inhibitory signals and down-regulation of the immune response. CD85J / LILRB1 interaction with HLA-B or HLA-E leads to inhibition of the signal triggered by FCER1A and inhibits serotonin release. CD85J / LILRB1 inhibits FCGR1A-mediated phosphorylation of cellular proteins and mobilization of intracellular calcium ions.

This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 75.6 kDa. As a result of glycosylation, the protein migrates as 100-116 kDa under reducing (R) condition, and 150 kDa under non
Spécification

Spécification

AAH15731.1
HEK293
Human
75.6 kDa
200 µg
Gly 24 - His 458
CD85J,LILRB1,CD85,ILT2,LIR1,MIR7
Precision Avi
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>95% as determined by SDS-PAGE
Biotinylated Human LILRB1, Fc,Avitag (CDJ-H82F7) is expressed from human 293 cells (HEK293). It contains AA Gly 24 - His 458 (Accession # D9IDM8-1).
-20°C
LILRB1
Fc Tag and Avi Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions