missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human OX40 Protein, Fc, Avitag™

Biotinylated Protein

Marque:  ACROBiosystems OX0-H82F7-200UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877397

  • 1392.13€ / 200µg
Date d’expédition estimée: 04-01-2022

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Tumor necrosis factor receptor superfamily member 4 (TNFRSF4) is also known as ACT35 antigen, OX40L receptor, TAX transcriptionally-activated glycoprotein 1 receptor, CD antigen CD134, OX40. OX40 / TNFRSF4 contains four TNFR-Cys repeats. TNFRSF4 is receptor for TNFSF4 / OX40L / GP34 and can interacts with TRAF2, TRAF3 and TRAF5.

This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 48.6 kDa. The protein migrates as 60-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Spécification

Spécification

NP_003318
HEK293
Human
48.6 kDa
200 µg
Leu 29 - Ala 216
TNFRSF4,OX40,CD134,OX40L receptor,ACT35,TXGP1L
Precision Avi
Tris with Glycine, Arginine and NaCl, pH7.5
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>95% as determined by SDS-PAGE
Biotinylated Human O x 40, Fc,Avitag (O x 0-H82F7) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Ala 216 (Accession # NP_003318).
-20°C
OX40
Fc Tag and Avi Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions