missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human TNF-alpha Protein, epitope tag free, ultra sensitivity (primary amine labeling) (active trimer)

MALS verified

Marque:  ACROBiosystems TNA-H8211-200UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15867837

  • 1387.09€ / 200µg
Date d’expédition estimée: 28-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Tumor necrosis factor alpha (TNFα) is a cytokine produced primarily by monocytes and macrophages. It is found in synovial cells and macrophages in the tissues.The primary role of TNFα is in the regulation of immune cells. TNFα is able to induce apoptotic cell death, to induce inflammation, and to inhibit tumorigenesis and viral replication. Dysregulation of TNFα production has been implicated in a variety of human diseases, including major depression, Alzheimer's disease and cancer. Recombinant TNFα is used as an immunostimulant under the INN tasonermin. TNFα can be produced ectopically in the setting of malignancy and parallels parathyroid hormone both in causing secondary hypercalcemia and in the cancers with which excessive production is associated.

The product does NOT contain any epitope tags. The protein has a calculated MW of 17.4 kDa. The protein migrates as 18 kDa on a SDS-PAGE gel under reducing (R) condition.
Spécification

Spécification

NP_000585.2
HEK293
Human
17.4 kDa
200 µg
Val 77 - Leu 233
DIF,TNF-alpha,TNFA,TNFSF2,cachexin,cachectin,TNFα
UltraLys
PBS, pH7.4
Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
Powder
>95% as determined by reduced SDS-PAGE
MABSol™ Biotinylated Human TNF-alpha, epitope tag free, primary amine labeling (active trimer) (MALS verified) (TNA-H8211) is expressed from human HEK293 cells. It contains AA Val 77 - Leu 233 (Accession # NP_000585.2). It is the biotinylated form of ActiveMax™ Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (TNA-H4211).
-20°C
TNF-alpha
Tag Free
Vidéos
FDS
Documents
Certificats
Promotions

Promotions