missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Biotinylated Human VEGF R3 / FLT4 Protein, His, Avitag™
Biotinylated Protein
423.38€ - 1681.42€
Spécification
Numéro d’adhésion | NP_002011.2 |
---|---|
Poids moléculaire | 88.2 kDa |
Source | Biotinylated Human VEGF R3, His,Avitag (FL4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Accession # P35916-1). |
Immunogène | Tyr 25 - Ile 776 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15886767
|
ACROBiosystems
FL4-H82E1-25UG |
25 μg |
423.38€
25µg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15876767
|
ACROBiosystems
FL4-H82E1-200UG |
200 μg |
1681.42€
200µg |
Expédition estimée: 07-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Vascular endothelial growth factor receptor 3 (VEGF R3), also known as FLT-4, together with the other two members VEGFR1 (FLT-1) and VEGFR2 (KDR/Flk-1) are receptors for vascular endothelial growth factors (VEGF) and belong to the class III subfamily of receptor tyrosine kinases (RTKs). VEGF R3 mediates lymphangiogenesis in response to VEGF-C and VEGF-D. VEGF R3 is widely expressed in the early embryo but becomes restricted to lymphatic endothelia at later stages of development. It is likely that VEGF R3 may be important for lymph angiogenesis.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 88.2 kDa. The protein migrates as 115-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_002011.2 | |
Biotinylated Human VEGF R3, His,Avitag (FL4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Tyr 25 - Ile 776 (Accession # P35916-1). | |
-20°C | |
VEGF R3 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
88.2 kDa | |
Tyr 25 - Ile 776 | |
Flt-4,FLT4,LMPH1A,PCLFLT41,VEGFR3,VEGFR-3,FLT-4,FLT-41,FLT41,PCL | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |