Learn More
ACROBiosystems Biotinylated Influenza A [A/Hong Kong/483/97(H5N1)] Hemagglutinin 1 (HA1) Protein, Avitag™,His Tag
Biotinylated Protein
sur devis
Spécification
Numéro d’adhésion | AAC32099.1 |
---|---|
Poids moléculaire | 39.6 kDa |
Source | Biotinylated Influenza A [A/Hong Kong/483/97(H5N1)] HA1, Avitag,His Tag (HA1-V82E4) is expressed from human 293 cells (HEK293). It contains AA Asp 17 - Arg 339 (Accession # AAC32099.1). |
Immunogène | Asp 17 - Arg 339 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15866907
|
ACROBiosystems
HA1-V82E4-200UG |
200 µg |
sur devis
200µg |
N/A | Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Cet article nécessite une mise à jour du prix avant d'être commandé. Demandez un prix pour cet article ici. |
|||||||||
15876907
|
ACROBiosystems
HA1-V82E4-25UG |
25 µg |
sur devis
25µg |
N/A | Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Cet article nécessite une mise à jour du prix avant d'être commandé. Demandez un prix pour cet article ici. |
|||||||||
Description
Influenza, commonly known as “the flu”, is an infectious disease of birds and mammals caused by RNA viruses of the family Orthomyxoviridae, the influenza viruses. The virus is divided into three main types (Influenzavirus A, Influenzavirus B, and Influenzavirus C), which are distinguished by differences in two major internal proteins (hemagglutinin (HA) and neuraminidase (NA), which are the most important targets for the immune system. The type A viruses are the most virulent human pathogens among the three influenza types and cause the most severe disease. The serotypes that have been confirmed in humans, ordered by the number of known human pandemic deaths, are: H1N1, H2N2, H3N2, H5N1, H7N7, H1N2, H9N2, H7N2, H7N3, H10N7, H7N9. H7N9 is a serotype of the species Influenzavirus A (avian influenza virus or bird flu virus). H7 normally circulates amongst avian populations with some variants known to occasionally infect humans. An H7N9 virus was first reported to have infected humans in 2013 in China.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 39.6 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE).Spécification
AAC32099.1 | |
Biotinylated Influenza A [A/Hong Kong/483/97(H5N1)] HA1, Avitag,His Tag (HA1-V82E4) is expressed from human 293 cells (HEK293). It contains AA Asp 17 - Arg 339 (Accession # AAC32099.1). | |
-20°C | |
Hemagglutinin 1 (HA1) | |
Influenza | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
39.6 kDa | |
Asp 17 - Arg 339 | |
HA1,Hemagglutinin 1 | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / µg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |