missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Recombinant Protein L Protein, His Tag, ultra sensitivity (primary amine labeling, long spacer)

Recombinant Protein

Marque:  ACROBiosystems RPL-P814R-500UG

Informations supplémentaires : Poids : 0.00001kg

 Afficher plus de versions de ce produit

Code produit. 15877617

  • 80.70€ / 500µg
Date d’expédition estimée: 20-12-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

Protein L was isolated from the surface of bacterial species Peptostreptococcus magnus and was found to bind Ig(IgG,IgM,IgA,IgE and IgD) through L chain interaction, from which the name was suggested. Despite this wide-ranging binding capability with respect to Ig classes, Protein L is not a universal immunoglobilin-binding protein. Binding of Protein L to immunoglobulins is restricted to those containing kappa light chains (i.e., k chain of the VL domain). In humans and mice, kappa (k) light chains predominate. The remaining immunoglobulins have lambda (l) light chains. The recombinant protein contains four immunoglobulin (Ig) binding domains (Bdomains) of the native protein. Besides antibody, protein L is also suitable for binding of a wide range of antibody fragments such as Fabs, single-chain variable fragments (scFv), and domain antibodies (Dabs).

This protein carries a polyhistidine tag at the N-terminus. The protein migrates as 50-60 kDa on a SDS-PAGE gel under reducing (R) condition. This protein can bind to VL-Kappa.
Spécification

Spécification

Less than 1.0 EU / µg by the LAL method.
MABSol™ Biotin Labeled Proteins
>95% as determined by reduced SDS-PAGE
Biotinylated Recombinant Protein L, His Tag, primary amine labeling, long spacer (RPL-P814R) is expressed from E.coli cells. It is the biotinylated form of Recombinant Protein L, His Tag (RPL-P3141).
RPL,Protein L
UltraLys
PBS, pH7.4
E.coli
Powder
500 µg
-20°C
Protein L
His Tag
Vidéos
FDS
Documents
Certificats
Promotions

Promotions