missing translation for 'onlineSavingsMsg'
Learn More

Bis-(4-hydroxy-1-naphthyl)phenylmethanol, Thermo Scientific™

Catalog Number p-69255
Click to view available options
Quantité:
25 g
Conditionnement:
Glass bottle
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
6948-88-5
C27H20O3
392.454
MFCD00003979
OUYLDJXFDLBCTQ-UHFFFAOYSA-N
bis 4-hydroxy-1-naphthyl phenylmethanol, 4,4'-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy-alpha-4-hydroxynaphthyl-alpha-phenylnaphthalene-1-methanol, 4-hydroxy-4-hydroxynaphthalen-1-yl-phenyl-methyl naphthalen-1-ol, bis 4-hydroxynaphthyl phenylmethan-1-ol, 4,4'-hydroxy phenyl methylene di 1-naphthol, 4,4-alpha-hydroxybenzylidene di-1-naphthol, bis-4-hydroxy-1 naphthyl-phenyl-methanol, 4,4-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy 4-hydroxynaphthalen-1-yl benzyl naphthalen-1-ol
81382
4-[hydroxy-(4-hydroxynaphthalen-1-yl)-phenylmethyl]naphthalen-1-ol
C1=CC=C(C=C1)C(C2=CC=C(C3=CC=CC=C32)O)(C4=CC=C(C5=CC=CC=C54)O)O
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
6948-88-5
C27H20O3
392.454
MFCD00003979
OUYLDJXFDLBCTQ-UHFFFAOYSA-N
bis 4-hydroxy-1-naphthyl phenylmethanol, 4,4'-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy-alpha-4-hydroxynaphthyl-alpha-phenylnaphthalene-1-methanol, 4-hydroxy-4-hydroxynaphthalen-1-yl-phenyl-methyl naphthalen-1-ol, bis 4-hydroxynaphthyl phenylmethan-1-ol, 4,4'-hydroxy phenyl methylene di 1-naphthol, 4,4-alpha-hydroxybenzylidene di-1-naphthol, bis-4-hydroxy-1 naphthyl-phenyl-methanol, 4,4-hydroxy phenyl methylene bis naphthalen-1-ol, 4-hydroxy 4-hydroxynaphthalen-1-yl benzyl naphthalen-1-ol
81382
4-[hydroxy-(4-hydroxynaphthalen-1-yl)-phenylmethyl]naphthalen-1-ol
C1=CC=C(C=C1)C(C2=CC=C(C3=CC=CC=C32)O)(C4=CC=C(C5=CC=CC=C54)O)O

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Bis-(4-hydroxy-1-naphthyl)phenylmethanol
Point de fusion 121.0°C to 125.0°C
Couleur Red to Brown
Spectre IR Authentic
Conditionnement Glass bottle
Quantité 25 g
Beilstein 06, IV, 7666
Poids de la formule 392.46
Forme physique Powder
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'