missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14596-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the POU-III class of neural transcription factors. The encoded protein is involved in neuronal differentiation and enhances the activation of corticotropin-releasing hormone regulated genes. Overexpression of this protein is associated with an increase in the proliferation of melanoma cells.Spécification
BRN2 | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20265, P31360, P56222 | |
Brain 2, Brn 2, BRN2, Oct 7, OCT7, Octamer binding protein 7, OTF 7, OTF7, POU class 3 homeobox 2, POU3F2, POUF3 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
18992, 29588, 5454 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
POU3F2 | |
Liquid | |
Pou3f2 | |
BRN2 Fusion Protein Ag6131 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Rat, Mouse |