missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTBD6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13776-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
Adapter protein for the CUL3 E3 ubiquitin-protein ligase complex. Involved in late neuronal development and muscle formation.Spécification
BTBD6 | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8K2J9, Q96KE9 | |
BDPL, BTB (POZ) domain containing 6, BTBD6, Lens BTB domain protein | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
399566, 690367, 90135 |
Immunoprecipitation, Western Blot | |
Unconjugated | |
BTBD6 | |
Liquid | |
Btbd6 | |
BTBD6 Fusion Protein Ag4766 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse, Rat |