missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ BUB1B (Human) Recombinant Protein

Human BUB1B full-length ORF ( AAH18739, 1 a.a. - 1050 a.a.) recombinant protein with GST-tag at N-terminal.

Marque:  Abnova™ H00000701-P01.25ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16122391

  • 467.00€ / 25µg

Expédition estimée
22-12-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

  • Sequence: MAAVKKEGGALSEAMSLEGDEWELSKENVQPLRQGRIMSTLQGALAQESACNNTLQQQKRAFEYEIRFYTGNDPLDVWDRYISWTEQNYPQGGKESNMSTLLERAVEALQGEKRYYSDPRFLNLWLKLGRLCNEPLDMYSYLHNQGIGVSLAQFYISWAEEYEARENFRKADAIFQEGIQQKAEPLERLQSQHRQFQARVSRQTLLALEKEEEEEVFESSVPQRSTLAELKSKGKKTARAPIIRVGGALKAPSQNRGLQNPFPQQMQNNSRITVFDENADEASTAELSKPTVQPWIAPPMPRAKENELQAGPWNTGRSLEHRPRGNTASLIAVPAVLPSFTPYVEETAQQPVMTPCKIEPSINHILSTRKPGKEEGDSLQRVQSHQQASEEKKEKMMYCKEKIYAGVGEFSFEEIRAEVFRKKLKEQREAELLTSAEKRAEMQKQIEEMEKKLKEIQTTQQERTGDQQEETMPTKETTKLQIASESQKIPGMTLSSSVCQVNCCARETSLAENIWQEQPHSKGPSVPFSIFDEFLLSEKKNKSPPADPPRVLAQRRPLAVLKTSESITSNEDVSPDVCDEFTGIEPLSEDAIITGFRNVTICPNPEDTCDFARAARFVSTPFHEIMSLKDLPSDPERLLPEEDLDVKTSEDQQTACGTIYSQTLSIKKLSPIIEDSREATHSSGFSGSSASVASTSSIKCLQIPEKLELTNETSENPTQSPWCSQYRRQLLKSLPELSASAELCIEDRPMPKLEIEKEIELGNEDYCIKREYLICEDYKLFWVAPRNSAELTVIKVSSQPVPWDFYINLKLKERLNEDFDHFCSCYQYQDGCIVWHQYINCFTLQDLLQHSEYITHEITVLIIYNLLTIVEMLHKAEIVHGDLSPRCLILRNRIHDPYDCNKNNQALKIVDFSYSVDLRVQLDVFTLSGFRTVQILEGQKILANCSSPYQVDLFGIADLAHLLLFKEHLQVFWDGSFWKLSQNISELKDGELWNKFFVRILNANDEATVSVLGELAAEMNGVFDTTFQSHLNKALWKVGKLTSPGALLFQ
Spécification

Spécification

AAH18739
budding uninhibited by benzimidazoles 1 homolog beta (yeast)
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BUB1B
GST
701
Wheat germ expression system
25 μg
BUB1beta, BUBR1, Bub1A, MAD3L, SSK1, hBUBR1
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Vidéos
FDS
Documents