missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C10orf28 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24967-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
C10orf28 | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q7Z5L2 | |
C10orf28, C10orf28 protein, GIDRP86, PSORT | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
27291 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
R3HCC1L | |
Liquid | |
R3HCC1L | |
C10orf28 Fusion Protein Ag21740 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |