missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C14orf28 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 26130-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Descripción
C14orf28 is a protein coding gene.Especificaciones
C14orf28 | |
Polyclonal | |
Unconjugated | |
C14orf28 | |
c14_5270, C14orf28, DRIP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
122525 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4W4Y0 | |
C14orf28 | |
C14orf28 Fusion Protein Ag23441 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |