Learn More
C17orf48 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marque: Invitrogen PA554740
Code nomenclature Nacres: NA.41
Informations supplémentaires : Poids : 0.02000kg
Description
Immunogen sequence: QSSPKYEQCM KILREHNPNT ELNSPQGLSE PQFVQFNGGF SQEQLNWLNE VLTFSDTNQE KVVIVSHLPI YP Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.
C17orf48 Polyclonal antibody specifically detects C17orf48 in Human samples. It is validated for Immunohistochemistry, Western Blot.Spécification
C17orf48 | |
Polyclonal | |
Unconjugated | |
ADPRM | |
2310004I24Rik; ADPRibase-Mn; ADP-ribose/CDP-alcohol diphosphatase, manganese dependent; ADP-ribose/CDP-alcohol diphosphatase, manganese-dependent; ADP-ribose/CDP-alcohol pyrophosphatase; Adprm; C17orf48; CDP-choline phosphohydrolase; liver manganese (II)-dependent ADP-ribose/CDP-alcohol pyrophosphatase; manganese-dependent ADP-ribose/CDP-alcohol diphosphatase; Manganese-dependent ADP-ribose/CDP-alcohol diphosphatase-like protein; MDS006; NBLA03831; RGD1309906 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
56985 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q3LIE5 | |
ADPRM | |
Recombinant protein corresponding to Human C17orf48 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only