missing translation for 'onlineSavingsMsg'
Learn More

C1D Recombinant Protein, Abnova

Marque:  Abnova H00010438-P02.10ug

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16125092

  • 310.00€ / 10µg

Expédition estimée
21-12-2020

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Spécification

Spécification

AAH05235
Solution
10438
C1D (Human) Recombinant Protein (P02)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C1D
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
41.25
8
Glutathione Sepharose 4 Fast Flow
10 ug
MAGEEINEDYPVEIHEYLSAFENSIGAVDEMLKTMMSVSRNELLQKLDPLEQAKVDLVSAYTLNSMFWVYLATQGVNPKEHPVKQELERIRVYMNRVKEITDKKKAGKLDRGAASRFVKNALWEPKSKNASKVANKGKSKS
MGC12261|MGC14659|SUNCOR
C1D
Wheat Germ (in vitro)
GST
Vidéos
FDS
Documents