missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf152 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24870-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
C20orf152 | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96M20, Q9D5U8 | |
C20orf152, CNBD2 | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
140894, 70873 |
Western Blot | |
Unconjugated | |
CNBD2 | |
Liquid | |
Cnbd2 | |
C20orf152 Fusion Protein Ag21668 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Human |