missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C21orf45 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 25832-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
C21orf45, also named as MIS18A, C21orf46 and FASP1, is required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis. C21orf45 has dimer (∽52 kDa) and heterodimer(∽105 kDa) forms.Spécification
C21orf45 | |
Polyclonal | |
Unconjugated | |
MIS18A | |
B28, C21orf45, C21orf46, FAPP1 associated protein 1, FASP1, MIS18A, MIS18alpha, Protein Mis18 alpha | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54069, 66578 | |
-20°C | |
Liquid |
Immunofluorescence, Western Blot, Immunocytochemistry | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9CZJ6, Q9NYP9 | |
MIS18A | |
C21orf45 Fusion Protein Ag22922 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |