missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C2orf67 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24183-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
C2orf67, also termed as chromosome 2 open reading frame 67, encodes a 705 amino-acid protein. It exists a variant, which encodes 987 amino-acid protein. The C2orf56 may express in lymphoid /immune and nervous system. The specific function of C2orf67 is still non-known.Spécification
C2orf67 | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A0AUZ9, Q5DTI6 | |
C2orf67, KANSL1L | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
151050, 68691 |
Immunohistochemistry (Paraffin), Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
KANSL1L | |
Liquid | |
Kansl1l | |
C2orf67 Fusion Protein Ag21471 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human, Mouse |