missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C3orf64 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27595-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
C3orf64 | |
Polyclonal | |
Unconjugated | |
EOGT | |
AER61, C3orf64, EGF domain-specific O-linked GlcNAc transferase, EOGT, EOGT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
101351, 285203 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5NDL2, Q8BYW9 | |
EOGT | |
C3orf64 Fusion Protein Ag25932 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |