missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C9orf11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 23168-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Spécification
C9orf11 | |
Polyclonal | |
Unconjugated | |
EQTN | |
Afaf, C9orf11, EQTN, Equatorin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54586 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot, Immunofluorescence | |
0.65 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9NQ60 | |
EQTN | |
C9orf11 Fusion Protein Ag19656 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |