missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CA5B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13342-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes responsible for catalyzing the reversible hydration of carbon dioxide.CAs show extensive diversity in their distribution and subcellular localization.They are involved in a variety of biological processes, including calcification, bone resorption, respiration, acid-base balance and the formation of aqueous humor, saliva, gastric juice and cerebrospinal fluid. CA VB, also known as carbonate dehydratase VB, is one of two isoforms of CA V. It localizes to the mitochondria and is involved in metabolic processes. CA VB is predomitly expressed in heart, pancreas, lung, placenta, kidney and skeletal muscle. It exhibits highest homology with family member CA VA (the second isoform of CA V); however, unlike CA VA, it is not expressed in the liver, suggesting that it plays a significantly different physiological role.Spécification
CA5B | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q66HG6, Q9QZA0, Q9Y2D0 | |
CA VB, CA5B, Carbonate dehydratase VB, Carbonic anhydrase VB | |
Rabbit | |
IgG | |
20 μL | |
-20°C | |
Primary | |
11238, 302669, 56078 |
Western Blot | |
Unconjugated | |
CA5B | |
Liquid | |
Ca5b | |
CA5B Fusion Protein Ag4166 | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Mouse, Rat, Human |