missing translation for 'onlineSavingsMsg'
Learn More

CABC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marque:  Proteintech 15528-1-AP-150UL

Informations supplémentaires : Poids : 0.00010kg

 Afficher plus de versions de ce produit

Code produit. 16804744

  • 329.85€ / 150µL

Expédition estimée
04-03-2021

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!


Description

Description

This gene encodes a mitochondrial protein similar to yeast ABC1, which functions in an electron-transferring membrane protein complex in the respiratory chain. It is not related to the family of ABC transporter proteins. Expression of this gene is induced by the tumor suppressor p53 and in response to DNA damage, and inhibiting its expression partially suppresses p53-induced apoptosis. Alternatively spliced transcript variants have been found; however, their full-length nature has not been determined.
Spécification

Spécification

CABC1
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5BJQ0, Q60936, Q8NI60
ADCK3, ARCA2, CABC1, Chaperone ABC1 like, COQ8, SCAR9
Rabbit
IgG
150 μL
-20°C
Primary
360887, 56997, 67426
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
COQ8A
Liquid
Coq8a
CABC1 Fusion Protein Ag7612
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Vidéos
FDS
Documents