missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNA1S Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22279-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
The antibody is specific to CACNA1S.
Voltage-sensitive calcium channels mediate the entry of calcium into many types of excitable cells and thus play a key role in neurotransmitter release and excitation-contraction (E-C) coupling. The 1,4-dihydropyridines (DHPs) are synthetic organic compounds which can be used to identify the L-type calcium channels that are found in all types of vertebrate muscle, neuronal and neuroendocrine cells. The DHP receptor is part of the L-type calcium channel complex and is thought to be the voltage sensor in E-C coupling. The purified DHP receptor isolated from triads is composed of at least four subunits. The alpha-1 subunit contains the binding site for the DHPs and shows high sequence homology to the voltage gated sodium channel. The alpha-2 subunit is a large glycoprotein associated with the DHP receptor which was first described in skeletal muscle and is also found in high concentrations in other excitable tissues such as cardiac muscle and brain and in low concentrations in most other tissues studied. The other two subunits that co-purify with the DHP receptor are termed beta and gamma.Spécification
CACNA1S | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13698 | |
CACH1, CACN1, CACNA1S, CACNL1A3, Cav1.1, CCHL1A3, HOKPP, hypoPP, MHS5 | |
Rabbit | |
IgG | |
150 μL | |
-20°C | |
Primary | |
779 |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
CACNA1S | |
Liquid | |
CACNA1S | |
Peptide | |
Antigen Affinity Chromatography | |
RUO | |
Antibody | |
Polyclonal | |
Human |