missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CACNB4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17770-1-AP-150UL
Informations supplémentaires : Poids : 0.00010kg
Description
This gene encodes a member of the beta subunit family of voltage-dependent calcium channel complex proteins. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. The protein encoded by this locus plays an important role in calcium channel function by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Certain mutations in this gene have been associated with idiopathic generalized epilepsy and juvenile myoclonic epilepsy. Multiple transcript variants encoding different isoforms have been found for this gene.Spécification
CACNB4 | |
Polyclonal | |
Unconjugated | |
CACNB4 | |
CAB4, CACNB4, CACNLB4, EA5, EJM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12298, 58942, 785 | |
-20°C | |
Liquid |
Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
D4A055, O00305, Q8R0S4 | |
Cacnb4 | |
CACNB4 Fusion Protein Ag12178 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |