missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cadherin-17 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24339-1-AP-20UL
Informations supplémentaires : Poids : 0.00010kg
Description
Cadherins are calcium dependent cell adhesion proteins. They preferentially interact with themselves in a homophilic manner in connecting cells; cadherins may thus contribute to the sorting of heterogeneous cell types.Spécification
Cadherin-17 | |
Polyclonal | |
Unconjugated | |
CDH17 | |
Cadherin-17, CDH16, CDH17, HPT 1, HPT1, LI cadherin, Liver intestine cadherin | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1015, 12557 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.41 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q12864, Q9R100 | |
CDH17 | |
Cadherin-17 Fusion Protein Ag19488 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |